AMBH93D53695 Ambeed, Inc.

(E)-4-(4-Methylpiperazin-1-yl)but-2-enoic acid hydrochloride

  • MDL number MFCD30067574
  • Cas Number : 1472802-56-4
  • Molecular Weight : 220.70

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,2-8°C
Physical Form solid
Shipping Temperature Normal
Country of Origin China
Product Link https://www.ambeed.com/products/1472802-56-4.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1472802-56-4.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart