AMBH93D536A2 Ambeed, Inc.

1-Propylpiperazine

  • MDL number MFCD00044591
  • Cas Number : 21867-64-1
  • Molecular Weight : 128.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Keep in dark place,Sealed in dry,Room Temperature
Physical Form Solid or liquid
Shipping Temperature Normal
Country of Origin China
Boiling point 176.7°C at 760 mmHg
Product Link https://www.ambeed.com/products/21867-64-1.html

Safety Information

signalword Warning
pcodes P264-P280-P302+P352-P305+P351+P338-P332+P313-P337+P313-P362
MSDS https://www.ambeed.com/products/21867-64-1.html
pictograms GHS07
hstatements H315-H319-H320
Availability details for
Quantity
  • Availability
  • Add to Cart