AMBH93D536A5 Ambeed, Inc.

cis-1-(tert-Butoxycarbonyl)-3-fluoropiperidine-4-carboxylic acid

  • MDL number MFCD28016232
  • Cas Number : 1628475-90-0
  • Molecular Weight : 247.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym rel-(3R,4S)-1-(tert-Butoxycarbonyl)-3-fluoropiperidine-4-carboxylic acid
Purity 97
Storage temp. Sealed in dry,2-8°C
Physical Form solid
Shipping Temperature Normal
Country of Origin China
Product Link https://www.ambeed.com/products/1628475-90-0.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1628475-90-0.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart