AMBH93D536A8 Ambeed, Inc.

2,2,2-Trifluoro-1-(1-methyl-1H-indol-3-yl)ethanol

  • MDL number MFCD02571216
  • Cas Number : 318-54-7
  • Molecular Weight : 229.20

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,2-8°C
Physical Form solid
Shipping Temperature Normal
Country of Origin China
Boiling point 299.8°C at 760 mmHg
Product Link https://www.ambeed.com/products/318-54-7.html

Safety Information

signalword Warning
pcodes P264-P280-P302+P352-P305+P351+P338-P332+P313-P337+P313-P362
MSDS https://www.ambeed.com/products/318-54-7.html
pictograms GHS07
hstatements H315-H319-H320
Availability details for
Quantity
  • Availability
  • Add to Cart