AMBH93D536AD Ambeed, Inc.

2-(4-Hydroxyphenyl)benzo[b]thiophen-6-ol

  • MDL number MFCD16619490
  • Cas Number : 63676-22-2
  • Molecular Weight : 242.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Keep in dark place,Inert atmosphere,Store in freezer, under -20°C
Physical Form solid
Shipping Temperature Normal
Country of Origin China
Product Link https://www.ambeed.com/products/63676-22-2.html

Safety Information

signalword Warning
pcodes P301+P312+P330
MSDS https://www.ambeed.com/products/63676-22-2.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart