AMBH93D536B1 Ambeed, Inc.

Diphenyl(pyren-1-yl)phosphine

  • MDL number MFCD00278755
  • Cas Number : 110231-30-6
  • Molecular Weight : 386.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,2-8°C
Physical Form solid
Shipping Temperature Normal
Country of Origin China
Boiling point 551.5°C at 760 mmHg
Product Link https://www.ambeed.com/products/110231-30-6.html

Safety Information

signalword Warning
pcodes P264-P280-P302+P352+P332+P313+P362+P364-P305+P351+P338+P337+P313
MSDS https://www.ambeed.com/products/110231-30-6.html
pictograms GHS07
hstatements H315-H319
Availability details for
Quantity
  • Availability
  • Add to Cart