AMBH93D536B7 Ambeed, Inc.

(4'-(tert-Butyl)-[1,1'-biphenyl]-2-yl)boronic acid

  • MDL number MFCD18383634
  • Cas Number : 1228458-46-5
  • Molecular Weight : 254.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,2-8°C
Physical Form solid
Shipping Temperature Normal
Country of Origin China
Product Link https://www.ambeed.com/products/1228458-46-5.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338-P304+P340-P405-P501
MSDS https://www.ambeed.com/products/1228458-46-5.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart