AMBH93D536BA Ambeed, Inc.

1-Methyl-4-(piperidin-4-yl)piperazine trihydrochloride

  • MDL number MFCD22573740
  • Cas Number : 1621519-77-4
  • Molecular Weight : 292.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin China
Product Link https://www.ambeed.com/products/1621519-77-4.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/1621519-77-4.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart