AMBH93D536BC Ambeed, Inc.

5-(2-Fluorophenyl)-5-oxopentanoic acid

  • MDL number MFCD01320116
  • Cas Number : 199664-70-5
  • Molecular Weight : 210.20

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin China
Product Link https://www.ambeed.com/products/199664-70-5.html

Safety Information

signalword Danger
pcodes P261-P264-P270-P271-P280-P302+P352-P304+P340-P310-P330-P361-P403+P233-P405-P501
MSDS https://www.ambeed.com/products/199664-70-5.html
pictograms GHS06
hstatements H301-H311-H331
Availability details for
Quantity
  • Availability
  • Add to Cart