AMBH93D536BD Ambeed, Inc.

3-Methylpyrazine-2-carbaldehyde

  • MDL number MFCD09834938
  • Cas Number : 25513-93-3
  • Molecular Weight : 122.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Flash Point 77.5°C
Storage temp. Inert atmosphere,2-8°C
Physical Form liquid
Shipping Temperature Normal
Country of Origin China
Boiling point 202.4°35.0°C at 760 mmHg
Product Link https://www.ambeed.com/products/25513-93-3.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/25513-93-3.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart