AMBH93D536BF Ambeed, Inc.

Benzyl ((3S,5R)-rel-5-(hydroxymethyl)pyrrolidin-3-yl)carbamate hydrochloride

  • MDL number N/A
  • Cas Number : 1217751-87-5
  • Molecular Weight : 286.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,2-8°C
Physical Form solid
Shipping Temperature Normal
Country of Origin China
Product Link https://www.ambeed.com/products/1217751-87-5.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1217751-87-5.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart