AMBH93D536C0 Ambeed, Inc.

N-Methyl-4-(dimethylamino)benzylamine Hydrochloride

  • MDL number MFCD07106762
  • Cas Number : 1158441-78-1
  • Molecular Weight : 200.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin China
Product Link https://www.ambeed.com/products/1158441-78-1.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1158441-78-1.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart