AMBH93D536C5 Ambeed, Inc.

6,8-Dichloropyrido[2,3-b]pyrazine

  • MDL number MFCD26402019
  • Cas Number : 1283075-60-4
  • Molecular Weight : 200.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,2-8°C
Physical Form Solid or liquid
Shipping Temperature Normal
Country of Origin China
Product Link https://www.ambeed.com/products/1283075-60-4.html

Safety Information

signalword Danger
pcodes P261-P264-P270-P271-P280-P302+P352-P304+P340-P310-P330-P361-P403+P233-P405-P501
MSDS https://www.ambeed.com/products/1283075-60-4.html
pictograms GHS06
hstatements H301-H311-H331
Availability details for
Quantity
  • Availability
  • Add to Cart