AMBH93D536C8 Ambeed, Inc.

4-(Trifluoromethyl)-1H-indazole

  • MDL number MFCD09263211
  • Cas Number : 1000339-98-9
  • Molecular Weight : 186.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,2-8°C
Physical Form solid
Shipping Temperature Normal
Country of Origin China
Product Link https://www.ambeed.com/products/1000339-98-9.html

Safety Information

signalword Warning
pcodes P261-P280-P301+P312-P302+P352-P305+P351+P338
MSDS https://www.ambeed.com/products/1000339-98-9.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart