AMBH93D536C9 Ambeed, Inc.

4-Bromo-3-chloro-2-fluoropyridine

  • MDL number MFCD11040607
  • Cas Number : 1017793-21-3
  • Molecular Weight : 210.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Inert atmosphere,2-8°C
Physical Form solid
Shipping Temperature Normal
Country of Origin China
Product Link https://www.ambeed.com/products/1017793-21-3.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1017793-21-3.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart