AMBH93D536CA Ambeed, Inc.

2-(Bromomethyl)-3-methoxypyridine

  • MDL number MFCD11520716
  • Cas Number : 889360-84-3
  • Molecular Weight : 202.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Flash Point 98.2°C
Storage temp. Sealed in dry,2-8°C
Physical Form solid
Shipping Temperature Normal
Country of Origin China
Boiling point 238.7°25.0°C at 760 mmHg
Product Link https://www.ambeed.com/products/889360-84-3.html

Safety Information

signalword Warning
pcodes P305+P351+P338
MSDS https://www.ambeed.com/products/889360-84-3.html
pictograms GHS07
hstatements H302-H319
Availability details for
Quantity
  • Availability
  • Add to Cart