AMBH93D536CB Ambeed, Inc.

3-Nitroquinolin-2-ol

  • MDL number MFCD09754108
  • Cas Number : 103029-75-0
  • Molecular Weight : 190.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,2-8°C
Physical Form Solid or liquid
Shipping Temperature Normal
Country of Origin China
Product Link https://www.ambeed.com/products/103029-75-0.html

Safety Information

signalword Warning
pcodes P264-P280-P302+P352-P305+P351+P338-P332+P313-P337+P313-P362
MSDS https://www.ambeed.com/products/103029-75-0.html
pictograms GHS07
hstatements H315-H319-H320
Availability details for
Quantity
  • Availability
  • Add to Cart