AMBH93D536CF Ambeed, Inc.

2-Amino-4-bromo-5-chlorophenol

  • MDL number MFCD25978069
  • Cas Number : 1037298-14-8
  • Molecular Weight : 222.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Sealed in dry,2-8°C
Physical Form solid
Shipping Temperature Normal
Country of Origin China
Product Link https://www.ambeed.com/products/1037298-14-8.html

Safety Information

signalword Danger
pcodes P261-P264-P270-P271-P272-P280-P285-P301+P312-P302+P352-P304+P340-P305+P351+P338-P330-P332+P313-P337+P313-P342+P311-P362-P403+P233-P405-P501
MSDS https://www.ambeed.com/products/1037298-14-8.html
pictograms GHS07,GHS08
hstatements H302-H315-H317-H319-H334-H335
Availability details for
Quantity
  • Availability
  • Add to Cart