AMBH93D536D0 Ambeed, Inc.

Methyl 1-methyl-1H-[1,2,4]triazole-3-carboxylate

  • MDL number MFCD13191558
  • Cas Number : 57031-66-0
  • Molecular Weight : 141.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Flash Point 113°C
Storage temp. Sealed in dry,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin China
Boiling point 263°C at 760 mmHg
Product Link https://www.ambeed.com/products/57031-66-0.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338
MSDS https://www.ambeed.com/products/57031-66-0.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart