AMBH93D536D1 Ambeed, Inc.

((1,3-Dioxolan-2-yl)methyl)tributylphosphonium bromide

  • MDL number MFCD09859807
  • Cas Number : 115754-62-6
  • Molecular Weight : 369.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Inert atmosphere,2-8°C
Physical Form solid
Shipping Temperature Normal
Country of Origin China
Product Link https://www.ambeed.com/products/115754-62-6.html

Safety Information

signalword Warning
pcodes P264-P280-P302+P352+P332+P313+P362+P364-P305+P351+P338+P337+P313
MSDS https://www.ambeed.com/products/115754-62-6.html
pictograms GHS07
hstatements H315-H319
Availability details for
Quantity
  • Availability
  • Add to Cart