AMBH93D536D9 Ambeed, Inc.

Msc2530818

  • Cas Number : 1883423-59-3
  • Molecular Weight : 340.81

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym (S)-(2-(4-Chlorophenyl)pyrrolidin-1-yl)(3-methyl-1H-pyrazolo[3,4-b]pyridin-5-yl)methanone
Purity 98
Storage temp. Sealed in dry,2-8°C
Physical Form solid
Shipping Temperature Normal
Country of Origin China
Product Link https://www.ambeed.com/products/1883423-59-3.html

Safety Information

signalword Warning
pcodes P261-P280-P301+P312-P302+P352-P305+P351+P338
MSDS https://www.ambeed.com/products/1883423-59-3.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart