AMBH93D536DD Ambeed, Inc.

Patentblueviolet

  • MDL number MFCD00867653
  • Cas Number : 68238-36-8
  • Molecular Weight : 566.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Sodium 2-((4-(diethylamino)phenyl)(4-(diethyliminio)cyclohexa-2,5-dien-1-ylidene)methyl)benzene-1,4-disulfonate
Purity 95
Storage temp. Inert atmosphere,2-8°C
Physical Form solid
Shipping Temperature Normal
Country of Origin China
Product Link https://www.ambeed.com/products/68238-36-8.html

Safety Information

signalword Warning
pcodes P261-P264-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P321-P332+P313-P337+P313-P362-P403+P233-P405-P501
MSDS https://www.ambeed.com/products/68238-36-8.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart