AMBH93D536E6 Ambeed, Inc.

4-Methoxyindolin-2-one

  • MDL number MFCD07368221
  • Cas Number : 7699-17-4
  • Molecular Weight : 163.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Flash Point 168.7°C
Storage temp. Sealed in dry,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin China
Boiling point 355.4°C at 760 mmHg
Product Link https://www.ambeed.com/products/7699-17-4.html

Safety Information

signalword Warning
pcodes P280
MSDS https://www.ambeed.com/products/7699-17-4.html
pictograms GHS07
hstatements H302-H317
Availability details for
Quantity
  • Availability
  • Add to Cart