AMBH93D536E7 Ambeed, Inc.

5-Ethynylpicolinaldehyde

  • MDL number MFCD18256079
  • Cas Number : 940911-03-5
  • Molecular Weight : 131.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Inert atmosphere,2-8°C
Physical Form solid
Shipping Temperature Shipped in cold pack
Country of Origin China
Product Link https://www.ambeed.com/products/940911-03-5.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338-P304+P340-P405-P501
MSDS https://www.ambeed.com/products/940911-03-5.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart