AMBH93D536E8 Ambeed, Inc.

(R)-5-((9H-Fluoren-9-yl)methoxy)-4-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-5-oxopentanoic acid

  • MDL number MFCD02094381
  • Cas Number : 252049-17-5
  • Molecular Weight : 547.60

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,2-8°C
Physical Form solid
Shipping Temperature Normal
Country of Origin China
Product Link https://www.ambeed.com/products/252049-17-5.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/252049-17-5.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart