AMBH93D536EC Ambeed, Inc.

5-Nitro-2-phenyl-1H-indole

  • MDL number MFCD00446220
  • Cas Number : 4993-87-7
  • Molecular Weight : 238.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Keep in dark place,Sealed in dry,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin China
Product Link https://www.ambeed.com/products/4993-87-7.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/4993-87-7.html
pictograms GHS07
hstatements H302-H317
Availability details for
Quantity
  • Availability
  • Add to Cart