AMBH93D536EE Ambeed, Inc.

3-Bromo-5-cyclopropyl-1,2,4-oxadiazole

  • MDL number MFCD12827917
  • Cas Number : 121562-08-1
  • Molecular Weight : 189.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,Store in freezer, under -20°C
Physical Form liquid
Shipping Temperature Shipped in cold pack
Country of Origin China
Product Link https://www.ambeed.com/products/121562-08-1.html

Safety Information

signalword Warning
pcodes P261-P264-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P330-P332+P313-P337+P313-P362-P403+P233-P405-P501
MSDS https://www.ambeed.com/products/121562-08-1.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart