AMBH93D536F1 Ambeed, Inc.

4-(Bromomethyl)-N,N-diphenylaniline

  • MDL number MFCD20485908
  • Cas Number : 183994-94-7
  • Molecular Weight : 338.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,2-8°C
Physical Form Solid or liquid
Shipping Temperature Normal
Country of Origin China
Product Link https://www.ambeed.com/products/183994-94-7.html

Safety Information

signalword Warning
pcodes P261-P264-P270-P271-P280-P301+P312+P330-P302+P352-P304+P340+P312-P305+P351+P338-P332+P313-P337+P313-P362-P403+P233-P405-P501
MSDS https://www.ambeed.com/products/183994-94-7.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart