AMBH93D536F4 Ambeed, Inc.

Isochroman-3-carboxylic acid

  • MDL number MFCD18416383
  • Cas Number : 1261578-13-5
  • Molecular Weight : 178.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin China
Product Link https://www.ambeed.com/products/1261578-13-5.html

Safety Information

signalword Warning
pcodes P261-P264-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P337+P313-P362-P403+P233-P405-P501
MSDS https://www.ambeed.com/products/1261578-13-5.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart