AMBH93D536F8 Ambeed, Inc.

(R)-1-(3,5-Dimethoxyphenyl)ethanamine hydrochloride

  • MDL number MFCD13191723
  • Cas Number : 1257106-72-1
  • Molecular Weight : 217.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Benzenemethanamine, 3,5-dimethoxy-alpha-methyl-, hydrochloride, (alphaR)-
Purity 95
Storage temp. Inert atmosphere,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin China
Product Link https://www.ambeed.com/products/1257106-72-1.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1257106-72-1.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart