AMBH93D5370E Ambeed, Inc.

4-(Dimethylamino)benzimidamide

  • MDL number MFCD05662967
  • Cas Number : 55978-60-4
  • Molecular Weight : 163.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Keep in dark place,Sealed in dry,2-8°C
Physical Form Solid or liquid
Shipping Temperature Normal
Country of Origin China
Boiling point 283.7°C at 760 mmHg
Product Link https://www.ambeed.com/products/55978-60-4.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/55978-60-4.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart