AMBH93D53714 Ambeed, Inc.

4-Bromo-3-methylpyridine hydrochloride

  • MDL number MFCD04966778
  • Cas Number : 40899-37-4
  • Molecular Weight : 208.48

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,2-8°C
Physical Form solid
Shipping Temperature Normal
Country of Origin China
Boiling point 250°C at 760 mmHg
Product Link https://www.ambeed.com/products/40899-37-4.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/40899-37-4.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart