AMBH93D53717 Ambeed, Inc.

5-(Trifluoromethyl)-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD11205085
  • Cas Number : 68744-64-9
  • Molecular Weight : 169.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym N/A
Purity 95
Flash Point 9.2°C
Storage temp. Inert atmosphere,2-8°C
Physical Form solid
Shipping Temperature Normal
Country of Origin China
Boiling point 91.7°C at 760 mmHg
Product Link https://www.ambeed.com/products/68744-64-9.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338
MSDS https://www.ambeed.com/products/68744-64-9.html
pictograms GHS07
hstatements H302-H312-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart