AMBH93D5371D Ambeed, Inc.

(2S,4S)-4-(((Benzyloxy)carbonyl)amino)pyrrolidine-2-carboxylic acid

  • MDL number MFCD08704526
  • Cas Number : 1279034-86-4
  • Molecular Weight : 264.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Keep in dark place,Sealed in dry,2-8°C
Physical Form solid
Shipping Temperature Normal
Country of Origin China
Product Link https://www.ambeed.com/products/1279034-86-4.html

Safety Information

signalword Warning
pcodes P261-P280-P301+P312-P302+P352-P305+P351+P338
MSDS https://www.ambeed.com/products/1279034-86-4.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart