AMBH93D53720 Ambeed, Inc.

(2S,4S)-1-Cbz-2-Hydroxymethyl-4-aminopyrrolidine hydrochloride

  • MDL number MFCD11101176
  • Cas Number : 1279038-54-8
  • Molecular Weight : 286.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,2-8°C
Physical Form solid
Shipping Temperature Normal
Country of Origin China
Product Link https://www.ambeed.com/products/1279038-54-8.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/1279038-54-8.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart