AMBH93D53723 Ambeed, Inc.

4-Methoxy-2-methylphenylacetic acid

  • MDL number MFCD02664778
  • Cas Number : 942-97-2
  • Molecular Weight : 180.20

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin China
Boiling point 327.8°C at 760 mmHg
Product Link https://www.ambeed.com/products/942-97-2.html

Safety Information

signalword Warning
pcodes P233-P260-P261-P264-P271-P280-P302+P352-P304-P304+P340-P305+P351+P338-P312-P321-P332+P313-P337+P313-P340-P362-P403-P403+P233-P405-P501
MSDS https://www.ambeed.com/products/942-97-2.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart