AMBH93D53725 Ambeed, Inc.

(R)-2-Amino-2-(thiophen-2-yl)acetic acid

  • MDL number MFCD00079612
  • Cas Number : 65058-23-3
  • Molecular Weight : 157.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Keep in dark place,Sealed in dry,2-8°C
Physical Form solid
Shipping Temperature Normal
Country of Origin China
Boiling point 311.8°C at 760 mmHg
Product Link https://www.ambeed.com/products/65058-23-3.html

Safety Information

signalword Warning
pcodes P261-P264-P270-P271-P280-P301+P312-P302+P352-P304+P340-P330-P363-P501
MSDS https://www.ambeed.com/products/65058-23-3.html
pictograms GHS07
hstatements H302-H312-H332
Availability details for
Quantity
  • Availability
  • Add to Cart