AMBH93D53730 Ambeed, Inc.

6-Deoxy-a-L-galactopyranose

  • MDL number MFCD00069812
  • Cas Number : 6696-41-9
  • Molecular Weight : 164.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 136-140°C at N/A mmHg
Synonym (2R,3S,4R,5S,6S)-6-Methyltetrahydro-2H-pyran-2,3,4,5-tetraol
Purity 95
Flash Point 149.7°C
Storage temp. Sealed in dry,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin China
Boiling point 323.9°C at 760 mmHg
Product Link https://www.ambeed.com/products/6696-41-9.html

Safety Information

signalword Warning
pcodes P233-P260-P261-P264-P271-P280-P302+P352-P304-P304+P340-P305+P351+P338-P312-P321-P332+P313-P337+P313-P340-P362-P403-P403+P233-P405-P501
MSDS https://www.ambeed.com/products/6696-41-9.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart