AMBH93D58E13 Ambeed, Inc.

(R)-1-Boc-3-bromopyrrolidine

  • MDL number MFCD17214727
  • Cas Number : 569660-97-5
  • Molecular Weight : 250.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Flash Point 122°C
Storage temp. Sealed in dry,Store in freezer, under -20°C
Physical Form solid
Shipping Temperature Shipped in cold pack
Country of Origin China
Boiling point 277.5°33.0°C at 760 mmHg
Product Link https://www.ambeed.com/products/569660-97-5.html

Safety Information

signalword Danger
pcodes P280-P373-P305+P351+P338-P301+P312-P337+P313-P501
MSDS https://www.ambeed.com/products/569660-97-5.html
pictograms GHS07,GHS09
hstatements H302-H319-H400
Availability details for
Quantity
  • Availability
  • Add to Cart