AMBH93D58E1C Ambeed, Inc.

3-Aminocrotononitrile

  • MDL number MFCD00008071
  • Cas Number : 1118-61-2
  • Molecular Weight : 82.10

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Keep in dark place,Inert atmosphere,2-8°C
Physical Form solid
Shipping Temperature Normal
Country of Origin China
Product Link https://www.ambeed.com/products/1118-61-2.html

Safety Information

signalword Danger
pcodes P261-P280-P302+P352
MSDS https://www.ambeed.com/products/1118-61-2.html
pictograms GHS06
hstatements H302-H311-H317
Availability details for
Quantity
  • Availability
  • Add to Cart