AMBH93D58E4D Ambeed, Inc.

(E)-3-(2-Chloro-4-fluorophenyl)acrylic acid

  • MDL number N/A
  • Cas Number : 174603-37-3
  • Molecular Weight : 200.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 245-249°C at N/A mmHg
Purity 98
Storage temp. Inert atmosphere,2-8°C
Physical Form solid
Shipping Temperature Normal
Country of Origin China
Product Link https://www.ambeed.com/products/174603-37-3.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338-P304+P340-P405-P501
MSDS https://www.ambeed.com/products/174603-37-3.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart