AMBH93D58E64 Ambeed, Inc.

Terutroban

  • MDL number MFCD28411833
  • Cas Number : 165538-40-9
  • Molecular Weight : 407.91

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym (R)-3-(6-(4-Chlorophenylsulfonamido)-2-methyl-5,6,7,8-tetrahydronaphthalen-1-yl)propanoic acid
Purity 98
Storage temp. Sealed in dry,Store in freezer, under -20°C
Physical Form solid
Shipping Temperature Normal
Country of Origin China
Product Link https://www.ambeed.com/products/165538-40-9.html

Safety Information

signalword Warning
pcodes P261-P264-P271-P280-P302+P352-P304+P340+P312-P305+P351+P338-P332+P313-P337+P313-P362-P403+P233-P405-P501
MSDS https://www.ambeed.com/products/165538-40-9.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart