AMBH93D58E67 Ambeed, Inc.

2,2-Bis[(4S)-4-benzyl-2-oxazolin-2-yl]propane

  • MDL number MFCD30343613
  • Cas Number : 176706-98-2
  • Molecular Weight : 362.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym (4S,4'S)-2,2'-(Propane-2,2-diyl)bis(4-benzyl-4,5-dihydrooxazole)
Purity 9899
Storage temp. Inert atmosphere,2-8°C
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/176706-98-2.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/176706-98-2.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart