AMBH93D58E70 Ambeed, Inc.

1,3,5-Trimethylcyclohexane

  • MDL number MFCD00042625
  • Cas Number : 1839-63-0
  • Molecular Weight : 126.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Sealed in dry,Room Temperature
Physical Form liquid
Shipping Temperature Normal
Country of Origin China
Product Link https://www.ambeed.com/products/1839-63-0.html

Safety Information

signalword Danger
pcodes P210-P233-P240-P241+P242+P243-P280-P303+P361+P353-P370+P378-P403+P235-P501
MSDS https://www.ambeed.com/products/1839-63-0.html
pictograms GHS02
hstatements H225
Availability details for
Quantity
  • Availability
  • Add to Cart