AMBH93D58E79 Ambeed, Inc.

Fmoc-�HoGlu(OtBu)-OH

  • MDL number MFCD01863053
  • Cas Number : 203854-49-3
  • Molecular Weight : 439.50

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 115-120°C
Synonym (S)-3-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-6-(tert-butoxy)-6-oxohexanoic acid
Purity 98
Storage temp. Sealed in dry,2-8°C
Physical Form solid
Shipping Temperature Normal
Country of Origin China
Boiling point 638.8°C at 760 mmHg
Product Link https://www.ambeed.com/products/203854-49-3.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/203854-49-3.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart