AMBH93D58E9C Ambeed, Inc.

(S)-5-Methoxy-1,2,3,4-tetrahydronaphthalen-2-amine hydrochloride

  • MDL number MFCD08443937
  • Cas Number : 58349-17-0
  • Molecular Weight : 213.70

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin China
Boiling point 295.7°C at 760 mmHg
Product Link https://www.ambeed.com/products/58349-17-0.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/58349-17-0.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart