AMBH93D58E9E Ambeed, Inc.

(E)-1-(3-(2-(7-Chloroquinolin-2-yl)vinyl)phenyl)prop-2-en-1-ol

  • MDL number MFCD09833496
  • Cas Number : 149968-10-5
  • Molecular Weight : 321.81

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 116-117.5°C
Purity 95
Storage temp. Sealed in dry,2-8°C
Physical Form solid
Shipping Temperature Normal
Country of Origin China
Boiling point 504.1°38.0°C at 760 mmHg
Product Link https://www.ambeed.com/products/149968-10-5.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/149968-10-5.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart