AMBH93D59134 Ambeed, Inc.

(S)-3-Amino-3-(naphthalen-1-yl)propanoic acid

  • MDL number MFCD04113670
  • Cas Number : 275826-46-5
  • Molecular Weight : 215.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Flash Point 203.2
Storage temp. Keep in dark place,Sealed in dry,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 412.3�t 760 mmHg
Product Link https://www.ambeed.com/products/275826-46-5.html

Safety Information

signalword Warning
pcodes P261-P280-P301+P312-P302+P352-P305+P351+P338
MSDS https://www.ambeed.com/products/275826-46-5.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart