AMBH93E4C188 Ambeed, Inc.

3-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-4,4,4-trifluorobutanoic acid

  • MDL number MFCD02682449
  • Cas Number : 194471-86-8
  • Molecular Weight : 379.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,2-8°C
Physical Form solid
Shipping Temperature Normal
Country of Origin China
Product Link https://www.ambeed.com/products/194471-86-8.html

Safety Information

signalword Warning
pcodes P261-P264-P270-P271-P280-P301+P312-P302+P352-P304+P340-P330-P363-P501
MSDS https://www.ambeed.com/products/194471-86-8.html
pictograms GHS07
hstatements H302-H312-H332
Availability details for
Quantity
  • Availability
  • Add to Cart