AOB640091149 AOBChem

4-Carboxy-3-fluorophenylboronic acid

  • MDL number MFCD01114671
  • Cas Number : 120153-08-4
  • Molecular Weight : 183.93

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 2-8*C
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin CN
Product Link https://aobchem.com/4-carboxy-3-fluorophenylboronic-acid.html

Safety Information

signalword Warning
pcodes P261 P305 P351 P338
MSDS https://aobchem.com/pub/media/files/sds/10055.pdf
pictograms GHS07
hstatements H315 H319 H335
Availability details for
Quantity
  • Availability
  • Add to Cart